Orders LinkContactUsLinkComponent
Synthware Round-Bottom Flasks with Three Necks, Angled
Synthware Round-Bottom Flasks with Three Necks, Angled
3000 ml, 45/50, 29/42
Catalog # 76264-092
Supplier:   Kemtech America
Supplier #:  F43583L
Synthware Round-Bottom Flasks with Three Necks, Angled
Catalog # 76264-092
Supplier:  Kemtech America
Synthware Round-Bottom Flasks with Three Necks, Angled
Synthware Round-Bottom Flasks with Three Necks, Angled
3000 ml, 45/50, 29/42
Price
$286.66
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents
Round bottom with [ST] joints.
  • 3 Angled necks
  • Ground glass joints
  • Heavy wall Borosilicate construction
  • Various capacities and joint sizes available
Heavy wall design. Hand-blown from tubing to ensure uniform wall thickness.
Specifications
  • Capacity:
    3000 ml
  • Center neck [ST] joint:
    45/50
  • Side neck ST joints:
    29/42