G-TRAP™ Empty FPLC Columns
PP, 1 ml
Catalog # 76263-822
Supplier: G-Biosciences
Supplier #: 786-1290
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
These empty columns are designed for fast protein liquid chromatography.
- Frit material is ultra-high molecular weight polyethylene
Columns are made of polypropylene and come in 1 ml and 5 ml sizes.
Delivery information: Includes column tube, sealing plug, sealing sleeve, top stop plug and bottom stop plug, top (large) frit, bottom (small) frit (inserted in column).
Specifications
- Capacity:1 ml
- Column hardware:PP
- Cat. No.:76263-822
- Max. Pressure:3 bar
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



