Hex Drive Bit Socket, Wiha Tools
Hex 3/8" Drive Bit Socket, 6", 1/4", 0.14 lbs.
Catalog # 76254-320
Supplier: WIHA TOOLS TE
Supplier #: 77179
Jump to:
Product Details & Documents
Socket feature easy bit replacement using standard set screw.
- Exact fit precision machined tip
- Impact resistant modified chrome-vanadium-molybdenum, tool steel
Specifications
- Description:Hex 3/8" Drive Bit Socket, 6"
- Size:1/4"
- Weight:0.14 lbs.
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



