Orders LinkContactUsLinkComponent
Adapter Assemblies, Apex Tools
Syringe Adapter Assembly, Plastic, 30 cc, 3/32" tube I.D., for use with Luer Lok™, Luer Slip™ and Tapered Tip Air Operated Syringes
  76253-452
Adapter Assemblies, Apex Tools
  76253-452
 :  
Adapter Assemblies, Apex Tools
Syringe Adapter Assembly, Plastic, 30 cc, 3/32" tube I.D., for use with Luer Lok™, Luer Slip™ and Tapered Tip Air Operated Syringes
Syringe adapter assembly have a unique design works with syringes.
  • Plugs directly into any Kahnetics shot meter
  • Swivel 360° on adapter head for ease of motion
Produces repeatable shots of material and requires minimal clean-up.
  • Description:
    Syringe Adapter Assembly, Plastic, 30 cc, 3/32" tube I.D., for use with Luer Lok™, Luer Slip™ and Tapered Tip Air Operated Syringes