CFP Boxes, Conductive Containers
CFP Box to hold 3 Trays
Catalog # 76252-580
Supplier: CONDUCTIVE CONTAINERS INC. TE
Supplier #: 08M-9200
Jump to:
Product Details & Documents
Conductive fluted plastic (CFP) containers with winged partitions and two strips of hooks and velcro on bottoms.
- Thickness of 3 mm
Specifications
- Description:CFP Box to hold 3 Trays