Coils
Pomona Minigrab® to banana cord, Yellow, 91 cm (3 ft.)
Catalog # 76250-646
Supplier: TESTEQUITY LLC TE
Supplier #: 3782-36-4
Jump to:
Product Details & Documents
Specifications
- Color:Yellow
- Description:Pomona Minigrab® to banana cord
- Length:91 cm (3 ft.)



