Order Entry
United States
Orders LinkContactUsLinkComponent
Mop Frames, TOUGH GUY, Grainger

Mop Frames, TOUGH GUY, Grainger

Snap-On Dust Mop Frame, 5", 24"
Catalog # 76249-708
Supplier:   GRAINGER
Supplier #:  1TZG3
Mop Frames, TOUGH GUY, Grainger
Catalog # 76249-708
Supplier:  GRAINGER
Mop Frames, TOUGH GUY, Grainger

Snap-On Dust Mop Frame, 5", 24"

Price
$29.55
Log in for availability
Each
Product Details & Documents

Dust mop frames replace worn or damaged frames or pair with compatible handles and heads to create a custom dust mop system.

  • Made from metal
  • Snap-on connection
Specifications
  • Description:
    Snap-On Dust Mop Frame
  • Depth:
    5"
  • Width:
    24"
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.