Order Entry
United States
Orders LinkContactUsLinkComponent
Cleanroom Tables with Solid Tabletop, Eagle Group

Cleanroom Tables with Solid Tabletop, Eagle Group

Cleanroom tables, square, Stainless steel, Stainless steel, 24×24", 13.6 kg
Catalog # 76247-674
Supplier:   EAGLE GROUP TE
Supplier #:  CRT2424T-H
Cleanroom Tables with Solid Tabletop, Eagle Group
Catalog # 76247-674
Supplier:  EAGLE GROUP TE
Cleanroom Tables with Solid Tabletop, Eagle Group

Cleanroom tables, square, Stainless steel, Stainless steel, 24×24", 13.6 kg

Price
$3,039.44
Log in for availability
Each
Product Details & Documents

Cleanroom tables made of a heavy gauge type 304 all stainless steel construction.

  • 14 gauge, electropolished or brushed stainless steel and reinforced with heavy gauge hat channels.
  • Legs and crossbracing are 16 gauge with welded crossbracing front to back and choice of center or rear knocked-down crossbracing
  • Alcohol-wiped and cleanroom bagged

For solid top tables, undershelves are heavy gauge construction with hat channel reinforcement on tables 84" (2134 mm) and longer.

 : Tables supplied with 1" (25 mm) adjustable stainless steel bullet feet and also provided with stainless steel assembly hardware and are shipped knock-down.

Specifications
  • Base/leg material:
    Stainless steel
  • Worksurface material:
    Stainless steel
  • Description:
    Cleanroom tables, square
  • W×D:
    24×24"
  • Weight:
    13.6 kg
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.