Order Entry
United States
Orders LinkContactUsLinkComponent
Pliers, Apex

Pliers, Apex

Jaw Kit, with 2 Pins, 2 Pads
Catalog # 76244-912
Supplier:   Cooper Tools
Supplier #:  52910KITN
Pliers, Apex
Catalog # 76244-912
Supplier:  Cooper Tools
Pliers, Apex

Jaw Kit, with 2 Pins, 2 Pads

Price
$34.52
Log in for availability
Each
Product Details & Documents

These unique compound action pliers deliver 40% reduced effort to cut.

Specifications
  • Description:
    Jaw Kit, with 2 Pins, 2 Pads
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.