Order Entry
United States
Orders LinkContactUsLinkComponent
Forceps - Serrated Tips

Forceps - Serrated Tips

Forceps with straight serrated tips, 6" (150 mm)
Catalog # 76243-828
Supplier:   Aven Tools
Supplier #:  18435
Forceps - Serrated Tips
Catalog # 76243-828
Supplier:  Aven Tools
Forceps - Serrated Tips
Forceps - Serrated Tips

Forceps with straight serrated tips, 6" (150 mm)

Price
$16.23
Log in for availability
Each Pack of 10
Product Details & Documents

General forceps with serrated tips.

  • Stainless steel finish provides excellent corrosion resistance

    Ideal for bending, separating and removing small items

    Long, straight tip is helpful when working in confined spaces

Specifications
  • Description:
    Forceps with straight serrated tips
  • Length:
    6" (150 mm)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.