Order Entry
United States
Orders LinkContactUsLinkComponent
Scribers, TESTEQUITY LLC TE

Scribers, TESTEQUITY LLC TE

7.56, 7.25, 7.56, 7.62", Straight scriber, hook scriber, offset angle scriber, 90° (3") scriber
Catalog # 76243-236
Supplier:   TESTEQUITY LLC TE
Supplier #:  58-0223
Scribers, TESTEQUITY LLC TE
Catalog # 76243-236
Supplier:  TESTEQUITY LLC TE
Scribers, TESTEQUITY LLC TE

7.56, 7.25, 7.56, 7.62", Straight scriber, hook scriber, offset angle scriber, 90° (3") scriber

Price
$19.15
Log in for availability
Each
Product Details & Documents
Specifications
  • Length:
    7.56, 7.25, 7.56, 7.62"
  • Includes:
    Straight scriber, hook scriber, offset angle scriber, 90° (3") scriber
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.