Minigrabber® Test Clip Patch Cords, Jensen Tools
Minigrabber® test clip patch cord, Red, 61 cm (2 ft.)
Catalog # 76242-826
Supplier: TESTEQUITY LLC TE
Supplier #: 4613-24-2
Jump to:
Product Details & Documents
Specifications
- Color:Red
- Description:Minigrabber® test clip patch cord
- Length:61 cm (2 ft.)
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



