Minigrabber® Test Clip Patch Cords, Jensen Tools
Minigrabber® test clip patch cord, Black, 30 cm (1 ft.)
Catalog # 76242-642
Supplier: TESTEQUITY LLC TE
Supplier #: 4613-12-0
Jump to:
Product Details & Documents
Specifications
- Color:Black
- Description:Minigrabber® test clip patch cord
- Length:30 cm (1 ft.)



