Minigrabber® Patch Cords, Jensen Tools
Patch cord, Minigrabber® to multi-stacking banana plug, Black, 30 cm (1 ft.)
Catalog # 76242-638
Supplier: TESTEQUITY LLC TE
: 3782-12-0
Jump to:
Product Details & Documents
Specifications
- Color:Black
- Description:Patch cord, Minigrabber® to multi-stacking banana plug
- Length:30 cm (1 ft.)



