Order Entry
United States
Orders LinkContactUsLinkComponent
Pliers, Apex

Pliers, Apex

Needle Nose Plier with Inside-Serrated Jaws, 5.7", Smooth and rounded
Catalog # 76242-488
Supplier:   Cooper Tools
Supplier #:  2411PD
Pliers, Apex
Catalog # 76242-488
Supplier:  Cooper Tools
Pliers, Apex

Needle Nose Plier with Inside-Serrated Jaws, 5.7", Smooth and rounded

Price
$283.26
Log in for availability
Each
Product Details & Documents

These unique compound action pliers deliver 40% reduced effort to cut.

Specifications
  • Description:
    Needle Nose Plier with Inside-Serrated Jaws
  • Length:
    5.7"
  • Jaw type:
    Smooth and rounded
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.