Order Entry
United States
Orders LinkContactUsLinkComponent
Xcelite® Hex Socket Type Nutdriver, Apex

Xcelite® Hex Socket Type Nutdriver, Apex

Nutdriver with Chrome Finish, Hollow Shaft, 5.55 mm (⁹⁄₃₂"), 79 mm (3⅛"), 184.2 mm (7.25")
Catalog # 76242-426
Supplier:   Cooper Tools
Supplier #:  THS9N
Xcelite® Hex Socket Type Nutdriver, Apex
Catalog # 76242-426
Supplier:  Cooper Tools
Xcelite® Hex Socket Type Nutdriver, Apex

Nutdriver with Chrome Finish, Hollow Shaft, 5.55 mm (⁹⁄₃₂"), 79 mm (3⅛"), 184.2 mm (7.25")

Price
$50.78
Log in for availability
Each
Product Details & Documents
Specifications
  • Description:
    Nutdriver with Chrome Finish, Hollow Shaft
  • Blade length:
    79 mm (3⅛")
  • Blade diameter:
    5.55 mm (⁹⁄₃₂")
  • Overall length:
    184.2 mm (7.25")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.