Order Entry
United States
Orders LinkContactUsLinkComponent
Pliers, Apex

Pliers, Apex

Thin Long Nose Plier, 5.5", Fine serrated
Catalog # 76242-058
Supplier:   Cooper Tools
Supplier #:  LN55VN
Pliers, Apex
Catalog # 76242-058
Supplier:  Cooper Tools
Pliers, Apex

Thin Long Nose Plier, 5.5", Fine serrated

Price
$151.57
Log in for availability
Each
Product Details & Documents

These unique compound action pliers deliver 40% reduced effort to cut.

Specifications
  • Description:
    Thin Long Nose Plier
  • Length:
    5.5"
  • Jaw type:
    Fine serrated