Order Entry
United States
Orders LinkContactUsLinkComponent
 

Xcelite® Hex Socket Type Nutdriver, Apex

Nutdriver with Chrome Finish, Hollow Shaft, 6.35 mm (¼"), 79 mm (3⅛"), 184.2 mm (7.25")
Catalog # 76241-474
Supplier:   Cooper Tools
 :  THS8N
Xcelite® Hex Socket Type Nutdriver, Apex
Catalog # 76241-474
Supplier:  Cooper Tools
Xcelite® Hex Socket Type Nutdriver, Apex

Nutdriver with Chrome Finish, Hollow Shaft, 6.35 mm (¼"), 79 mm (3⅛"), 184.2 mm (7.25")

Price
$59.98
Log in for availability
Each
Product Details & Documents
Specifications
  • Description:
    Nutdriver with Chrome Finish, Hollow Shaft
  • Blade length:
    79 mm (3⅛")
  • Blade diameter:
    6.35 mm (¼")
  • Overall length:
    184.2 mm (7.25")