Order Entry
United States
Orders LinkContactUsLinkComponent
Cleanroom Loop Hangers, Eagle Group

Cleanroom Loop Hangers, Eagle Group

Open loop cleanroom hangers, Chrome, 0.2 kg (0.4 lbs.)
Catalog # 76239-026
Supplier:   EAGLE GROUP TE
Supplier #:  OLH-C
Cleanroom Loop Hangers, Eagle Group
Catalog # 76239-026
Supplier:  EAGLE GROUP TE
Cleanroom Loop Hangers, Eagle Group

Open loop cleanroom hangers, Chrome, 0.2 kg (0.4 lbs.)

Price
$32.02
Log in for availability
Each
Product Details & Documents

It is used for freestanding or wall mounted gowning racks, removable and non-removable hanger models.

  • Chrome finish

Closed loop hangers discourage removal from the gowning area.

Specifications
  • Finish:
    Chrome
  • Description:
    Open loop cleanroom hangers
  • Weight:
    0.2 kg (0.4 lbs.)