Order Entry
United States
Orders LinkContactUsLinkComponent
Cleanroom Loop Hangers, Eagle Group

Cleanroom Loop Hangers, Eagle Group

Closed loop cleanroom hangers, Chrome, 0.2 kg (0.4 lbs.)
Catalog # 76238-536
Supplier:   EAGLE GROUP TE
Supplier #:  CLH-C
Cleanroom Loop Hangers, Eagle Group
Catalog # 76238-536
Supplier:  EAGLE GROUP TE
Cleanroom Loop Hangers, Eagle Group

Closed loop cleanroom hangers, Chrome, 0.2 kg (0.4 lbs.)

Price
$31.57
Log in for availability
Each
Product Details & Documents

It is used for freestanding or wall mounted gowning racks, removable and non-removable hanger models.

  • Chrome finish

Closed loop hangers discourage removal from the gowning area.

Specifications
  • Finish:
    Chrome
  • Description:
    Closed loop cleanroom hangers
  • Weight:
    0.2 kg (0.4 lbs.)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.