Dispensing Tips, EFD
Flexible dispensing tip, 22, —, Clear
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
22 GA TAPER TIP NEEDLE 50/PKG Catalog #37695-148
Product 37695-148 is similar to 76238-230 but manufacturer and specifications may vary.
Product Details & Documents
EFD produces the highest quality dispensing tips engineered hub flats for easy twist on, twist off. 360° SafetyLok™ thread ensures safe, positive attachment to syringe barrel.
- Tapered Smooth flow for application of medium
- Flexible Polypropylene shafts reach into hard-to-access areas
- Free of flash, burrs and contaminants
- Package labels include lot numbers for process control and traceability
For applications in chamfered, ESD-safe, PTFE-coated and PTFE-lined, microdot tips and oval tips.
Specifications
- Ø int.:—
- Color:Clear
- Description:Flexible dispensing tip
- Gauge:22