Orders LinkContactUsLinkComponent
Dispensing Tips, EFD INC.
Dispensing Tips, EFD INC.
18, Standard, Green
Catalog # 76238-224
Supplier #:  7018158
Dispensing Tips, EFD INC.
Catalog # 76238-224
Dispensing Tips, EFD INC.
18, Standard, Green
Price
$108.62
Log in for availability
Box of 50
Product Details & Documents
Smooth flow for application of medium- to high-viscosity fluids especially thick or particle-filled materials like epoxies, RTVs and braze pastes.
  • Free of flash, burrs and contaminants
  • Package labels include lot numbers for process control and traceability
  • Consistent from style to style and lot to lot
  • 360° SafetyLok™ thread ensures safe, positive attachment to syringe barrel
  • Engineered hub flats for easy twist on, twist off
Molded of polyethylene with UV-light block additive.
Packaging: Packaged 50 tips per box.
Specifications
  • Color:
    Green
  • Point style:
    Standard
  • Gauge:
    18