Order Entry
United States
Orders LinkContactUsLinkComponent
 

FectoCHO® Expression System

FectoCHO™ Expression System, 5×1 mL (Transfection Reagent), 6×1 L (Expression Medium), 5×1 ml
Catalog # 76234-738
FectoCHO® Expression System
Catalog # 76234-738
Supplier:  
CAS Number:  
FectoCHO® Expression System
FectoCHO® Expression System

FectoCHO™ Expression System, 5×1 mL (Transfection Reagent), 6×1 L (Expression Medium), 5×1 ml

CAS Number:  
Discontinued
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

FectoCHO® Expression system consists in the synergistic association of a novel CHO chemically defined medium, FectoCHO® CD Expression medium and a powerful transfection reagent, FectoPRO®.

  • Outstanding protein production yields in various CHO cells
  • Fast and effortless CHO cell adaptation
  • Chemically defined kit components
  • Excellent lot to lot reproducibility
  • Cost-effective solution

FectoCHO® CD Expression medium allows easy cultivation of various strains of CHO cells such as CHO-K1, CHO-S and ExpiCHO®-S , without the need of an extensive sequential adaptation. FectoPRO®-mediated transient transfection in this chemically defined medium leads to the highest protein and antibody yields ever reached using Transient Gene Expression, up to 4 times higher than with the competitor's systems.

 : Since 2002, Polyplus-transfection® is ISO 9001 certified and since then this certification has been successfully renewed every three years.

 : FectoPRO® transfection reagent, FectoPRO® Booster and FectoCHO® CD Expression Medium can be purchased separately.

Specifications

Specifications

  • Description:
    FectoCHO™ Expression System
  • Size:
    5×1 mL (Transfection Reagent), 6×1 L (Expression Medium)
  • Buffer volume:
    5×1 ml
  • Cat. No.:
    76234-738
  • Bioproduction:
    Yes
  • Cell Specific:
    CHO

Specifications