FectoCHO® Expression System
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
FectoCHO® Expression system consists in the synergistic association of a novel CHO chemically defined medium, FectoCHO® CD Expression medium and a powerful transfection reagent, FectoPRO®.
- Outstanding protein production yields in various CHO cells
- Fast and effortless CHO cell adaptation
- Chemically defined kit components
- Excellent lot to lot reproducibility
- Cost-effective solution
FectoCHO® CD Expression medium allows easy cultivation of various strains of CHO cells such as CHO-K1, CHO-S and ExpiCHO®-S , without the need of an extensive sequential adaptation. FectoPRO®-mediated transient transfection in this chemically defined medium leads to the highest protein and antibody yields ever reached using Transient Gene Expression, up to 4 times higher than with the competitor's systems.
Certifications: Since 2002, Polyplus-transfection® is ISO 9001 certified and since then this certification has been successfully renewed every three years.
Ordering information: FectoPRO® transfection reagent, FectoPRO® Booster and FectoCHO® CD Expression Medium can be purchased separately.
Documents
Polyplus-transfection® Protein and mAb Production Enhanced Transient Protein Production in CHO.pdf
Product Literature
Poster FectoCHO Expression System.pdf
Application Notes
Specifications
Specifications
- Description:FectoCHO™ Expression System
- Size:1 mL (Transfection Reagent), 1 L (Expression Medium)
- Buffer volume:1 ml
- Cat. No.:76234-736
- Bioproduction:Yes
- Cell Specific:CHO



