Orders LinkContactUsLinkComponent
Fieldtex® AED/O<sub>2</sub> Backpack
Fieldtex® AED/O2 Backpack
AED/O₂ backpack, Red, 14×6×24"
Catalog # 470303-554
Supplier:   FIELDTEX PRODUCTS
Supplier #:  911-84550RD
Fieldtex® AED/O<sub>2</sub> Backpack
Catalog # 470303-554
Supplier:  FIELDTEX PRODUCTS
Fieldtex® AED/O<sub>2</sub> Backpack
Fieldtex® AED/O2 Backpack
AED/O₂ backpack, Red, 14×6×24"
Price
$330.60
Log in for availability
Each
Product Details & Documents
This FTX backpack is like having an E.R. at your fingertips.
  • Heavy duty hardware
  • Designed for long term durability
  • Bag only
To facilitate a rapid response we designed one outer pocket to hold your AED and one smaller pocket above for extra AED pads and accessories. Mid-Section of case unzips to reveal a full-length compartment for collars, splints or other flat items. Interior of case has dedicated hold-downs for a 'D' size oxygen tank. A bag- valve-mask, airways, nasal cannulas and oxygen masks. Four color-coded see- thru pouches inside carry your assorted trauma and airway supplies.
For durability we coated all areas of abuse with heavy gauge vinyl to prevent wear and tear. two molded carry handles on top and side of pack as well as integrated backpack straps that can be secured when not in use. A load bearing strap is sewn completely around the outer case distributing the weight and preventing traumatic detachment of the carry handles.
Heavy-duty zippers and hardware plus reinforced double-stitched seams make this American-made backpack one that you will rely on for every call.
Ordering information: This item is sold as a bag only, and does not include any other materials that may appear in the photographs.
Specifications
  • Color:
    Red
  • Description:
    AED/O₂ backpack
  • W×D×H:
    14×6×24"