Orders LinkContactUsLinkComponent
Swingsette™ Biopsy Processing/Embedding Cassette in Quickload™ Stacks, Simport Scientific
Swingsette™ Biopsy Processing/Embedding Cassette in Quickload™ Stacks, Simport Scientific
Gray
Catalog # 76214-372
Supplier:   Simport Scientific
Supplier #:  M518-9T
Swingsette™ Biopsy Processing/Embedding Cassette in Quickload™ Stacks, Simport Scientific
Catalog # 76214-372
Supplier:  Simport Scientific
Swingsette™ Biopsy Processing/Embedding Cassette in Quickload™ Stacks, Simport Scientific
Swingsette™ Biopsy Processing/Embedding Cassette in Quickload™ Stacks, Simport Scientific
Gray
Price
$791.60
Log in for availability
Case of 2,000
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Secure specimens during processing with the Swingsette™ Biopsy Processing/Embedding Cassette, in Quickload™ stacks.
  • In Quickload™ stacks
  • 45° anterior writing area
  • Easy to assemble hinged lids
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
Certifications: CE-IVD.
Packaging: Each case contains 50 Quickload™ stacks of 40 cassettes, lids packaged separately.
Specifications
  • Color:
    Gray