Orders LinkContactUsLinkComponent
ESD SMD Boxes, Transforming Technologies
ESD SMD Boxes, Transforming Technologies
Small, 16×12×15 mm (int)
Catalog # 76209-954
Supplier #:  SM0880
ESD SMD Boxes, Transforming Technologies
Catalog # 76209-954
ESD SMD Boxes, Transforming Technologies
ESD SMD Boxes, Transforming Technologies
Small, 16×12×15 mm (int)
Price
$474.00
Log in for availability
Pack of 100
Product Details & Documents
Static-safe SMD boxes with spring-loaded hinges are designed to interlock for easy organization and storage.
  • Ideal for engineering labs
  • Available with conductive black or clear metalized lids
SMD plastic boxes have a unique interlocking modular design. Spring-loaded lids have a stay-open feature for convenient loading and removal of parts. Surface resistivity of 10² to 10⁵ ohms/cm². They are ideal for engineering labs, kitting, in-process handling, and safe transport of static-sensitive components.
Certifications: Meets or exceeds requirements of ANSI ESD-S20.20
Specifications
  • Description:
    Small
  • Ext. dimensions:
    16×12×15 mm (int)