Orders LinkContactUsLinkComponent
ESD SMD Boxes, Transforming Technologies
Extra-large, 68×57×15 mm (int)
  76209-952
 :  SM0876
ESD SMD Boxes, Transforming Technologies
  76209-952
ESD SMD Boxes, Transforming Technologies
ESD SMD Boxes, Transforming Technologies
Extra-large, 68×57×15 mm (int)
$68.50
Pack of 10
Static-safe SMD boxes with spring-loaded hinges are designed to interlock for easy organization and storage.
  • Ideal for engineering labs
  • Available with conductive black or clear metalized lids
SMD plastic boxes have a unique interlocking modular design. Spring-loaded lids have a stay-open feature for convenient loading and removal of parts. Surface resistivity of 10² to 10⁵ ohms/cm². They are ideal for engineering labs, kitting, in-process handling, and safe transport of static-sensitive components.
 : Meets or exceeds requirements of ANSI ESD-S20.20
  • Description:
    Extra-large
  • Ext. dimensions:
    68×57×15 mm (int)