ESD SMD Boxes, Transforming Technologies
Large, 40×37×15 mm (int)
Catalog # 76209-950
Supplier: Transforming Technologies
Supplier #: SM0875
Jump to:
Product Details & Documents
Static-safe SMD boxes with spring-loaded hinges are designed to interlock for easy organization and storage.
- Ideal for engineering labs
- Available with conductive black or clear metalized lids
SMD plastic boxes have a unique interlocking modular design. Spring-loaded lids have a stay-open feature for convenient loading and removal of parts. Surface resistivity of 10² to 10⁵ ohms/cm². They are ideal for engineering labs, kitting, in-process handling, and safe transport of static-sensitive components.
Certifications: Meets or exceeds requirements of ANSI ESD-S20.20
Specifications
- Description:Large
- Ext. dimensions:40×37×15 mm (int)