AGONIST TRI-DAP NOD1 1MG
Catalog # MSPP-TLRLTDAP
Supplier: Invivogen
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Vacuum Filtration Systems, Standard Line
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Tissue-Tek® O.C.T. Compound, Sakura® Finetek
Supplier: Sakura Finetek
VWR® Disposable Serological Pipettes
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Premium Aluminum Foil
Supplier: VWR International
Parafilm® M Sealing Film
Supplier: Bemis Company, Inc.
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
VWR® Weighing Paper
Supplier: VWR International
VWR® Chemical-Resistant Laboratory Marker
Supplier: VWR International