Order Entry
United States
Orders LinkContactUsLinkComponent
pH Gradient Buffers

pH Gradient Buffers

5.6 - 10.2
Catalog # 10053-825
pH Gradient Buffers
Catalog # 10053-825
Supplier:  
CAS Number:  
pH Gradient Buffers

5.6 - 10.2

CAS Number:  
Discontinued
Product Details & Documents

Deliver robust, reproducible gradients with the Thermo Scientific pH buffer platform, which is applicable to a wide range of MAbs and ready to use with existing LC columns and systems, without the need to formulate mobile phases.

  • Proprietary buffer formulations enable fast, robust and reproducible pH gradients that are simple to optimise and easily automated
  • Ready to use with existing LC columns and systems, without the need for time-consuming mobile phase adjustments
  • Applicable to the majority of MAbs

The pH gradient platform saves time in method development and facilitates method transfer to QA/QC for a wide range of MAb charge variants through a generic LC-based approach to MAb characterization.

All that is required to generate a pH gradient is a 1:10 dilution of the pH buffer concentrates. A linear pH gradient from pH 5.6 to 10.2 can simply be delivered by running a pump gradient from 100% eluent A to 100% eluent B.

Specifications

Specifications

  • pH:
    5.6 - 10.2

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.