Multiple Affinity Removal Column - Mouse 3, Agilent Technologies
Multiple Affinity Removal Column - Ms-3 (Mouse), 4.60 mm, 100 mm
Catalog # 76204-856
Jump to:
Discontinued
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
The Multiple Affinity Removal LC Column – Mouse 3 is designed as a simple way to remove THREE interfering high-abundant proteins from mouse plasma samples.
Removal of these abundant proteins improves the subsequent LC/MS and electrophoretic analysis of the serum sample by effectively expanding the dynamic range of the analysis.
Caution: For research use only.
Documents
Specifications
- Ø int.:4.60 mm
- Description:Multiple Affinity Removal Column - Ms-3 (Mouse)
- Length:100 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



