CELLS LCL 8664 RHESUS B-LYMPH. LYMPHOMA
Catalog # MSPP-CRL-1805
Supplier: 0000013421
CELLS LCL 8664 RHESUS B-LYMPH. LYMPHOMA
Catalog # MSPP-CRL-1805
Supplier: 0000013421
Supplier Number: CRL-1805
Frequently Bought Together
VWR® Cell Culture Dishes
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Serological Pipettes
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
Corning® Internal Thread Cryogenic Vials, Corning
Supplier: Corning
VWR® Disposable Petri Dishes
Supplier: VWR International
Parafilm® M Sealing Film
Supplier: Bemis Company, Inc.
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
VWR® Weighing Paper
Supplier: VWR International
VWR® Nitrile Examination Gloves, Black
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International
Reagent alcohol ACS (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International