Order Entry
United States
Orders LinkContactUsLinkComponent
DMNPE-caged Luciferin

DMNPE-caged Luciferin

Bottle, amber, 10 mg
Catalog # 103549-584
Marketsource
Supplier:   OZ Biosciences
Supplier #:  LC10000
DMNPE-caged Luciferin
Catalog # 103549-584
Supplier:  OZ Biosciences
Marketsource
CAS Number:  223920-67-0
DMNPE-caged Luciferin
DMNPE-caged Luciferin

Bottle, amber, 10 mg

CAS Number:  223920-67-0
Price
$219.97
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

The cell permeable DMNPE-caged Luciferin is as a bioluminescent luciferase substrate used to measure intracellular functions since it readily crosses cell membranes. It is a D-luciferin ester analogue of luciferin.

  • High purity
  • Cell permeable: efficient luciferin delivery into intact cells even at neutral pH
  • Ideal for in vivo applications
  • Induces prolonged release of luciferin by the action of cells’ esterases
  • Allows to follow changes in gene expression in live cells
  • Suitable for in vitro experiments

Once the caged luciferin is inside the cells, active luciferin can be released by the action of endogenous intracellular esterases or by a flash of UV light. This implies:
1. A more efficient delivery of luciferin into intact living cells & improved sensitivity analysis of in vivo luciferase assays
2. A long-term measurement of luciferase activity since hydrolysis by esterases allows a continuous supply of active Luciferin
DMNPE-caged luciferin appears a good alternative to D-Luciferin classically used to measure luciferase activity in live cells. Indeed, the difficulty to deliver D-Luciferin into living cells renders in vivo quantification limited to a certain extent- for example, permeability issues at neurtral pH.

Shipped at room temperature.
Store at -20°C and protect from light.

Synonyms:  D-Luciferin 1-(4,5-dimethoxy-2-nitrophenyl)ethyl ester
  • CAS Number:223920-67-0
  • Formula:C₂₁H₁₉N₃O₇S₂
  • Molecular Weight:489.52 g/mol
  •  :Packaged under nitrogen.
  • Storage Temperature:Freezer

Documents

Specifications

Specifications

  • Pack type:
    Bottle, amber
  • Size:
    10 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.