STEM CELLS HUMAN BONE MARROW-DERIVED MSC
Catalog # MSPP-PCS500012
Supplier: 0000013421
STEM CELLS HUMAN BONE MARROW-DERIVED MSC
Catalog # MSPP-PCS500012
Supplier: 0000013421
Supplier Number: PCS-500-012
Frequently Bought Together
Alconox® Powdered Precision Cleaners
Supplier: Alconox
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
Parafilm® M Sealing Film
Supplier: Bemis Company, Inc.
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
VWR® Weighing Paper
Supplier: VWR International
Corning® Internal Thread Cryogenic Vials, Corning
Supplier: Corning
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
2-Propanol ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Petri Dishes
Supplier: VWR International
Single-Use Needles, BD Medical
Supplier: BD
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Disposable Transfer Pipettes
Supplier: VWR International
VWR® Disposable Spatulas, PP
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International