GEL DNA RECOVERY KIT 10 PREPS F/LIGATION
Catalog # 76197-092
Supplier: Zymo Research
GEL DNA RECOVERY KIT 10 PREPS F/LIGATION
Catalog # 76197-092
Supplier: Zymo Research
Supplier Number: D4001T
Frequently Bought Together
VWR® Weighing Paper
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
Corning® Filter Top Tubes, Sterile, Corning
Supplier: Corning
VWR® Sterile 70% Isopropanol
Supplier: VWR International
Water, HiPerSolv CHROMANORM for HPLC, VWR Chemicals BDH®
Supplier: VWR International
Clorox® Concentrated Germicidal Bleach
Supplier: Janitorial Supplies
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Petri Dishes
Supplier: VWR International
Tissue-Tek® O.C.T. Compound, Sakura® Finetek
Supplier: Sakura Finetek
VWR® Universal Pipette Tips
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15×100 mm plate
Supplier: Hardy Diagnostics