SOIL MICROBE 96 MAGBEAD KIT QUICK-DNA
Catalog # 76197-080
Supplier: Zymo Research
SOIL MICROBE 96 MAGBEAD KIT QUICK-DNA
Catalog # 76197-080
Supplier: Zymo Research
Supplier Number: D6012-FM
Frequently Bought Together
VWR® Weighing Paper
Supplier: VWR International
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
VWR® Razor Blades
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Tissue-Tek® O.C.T. Compound, Sakura® Finetek
Supplier: Sakura Finetek
KIMTECH SCIENCE® KIMWIPES™ Delicate Task Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
VWR® Permanent Marking Pens
Supplier: VWR International