Laser Diode for Debye-Sears Effect, 3B Scientific
Green Laser Diode for Debye-Sears
Catalog # 470233-728
Supplier: American 3B Scientific
Supplier #: 1002579
Laser Diode for Debye-Sears Effect, 3B Scientific
Catalog # 470233-728
Supplier: American 3B Scientific
Jump to:
Product Details & Documents
Laser diode of protection classification II with 1 m connector lead and barrel connector for connecting ultrasonic cw generator.
- All lasers have been measured to determine their wavelength specifically and the results are logged
Laser diode of protection classification II with 1 m connector lead and barrel connector for connecting ultrasonic cw generator (U100061). Fits the laser holder of test vessel (U10008). All lasers have been measured to determine their wavelength specifically and the results are logged.
Specifications
- Description:Green Laser Diode for Debye-Sears