Order Entry
United States
Orders LinkContactUsLinkComponent
BUFFER AMV REVERSE TRANSCRIPTASE 400UL

BUFFER AMV REVERSE TRANSCRIPTASE 400UL

Catalog # 80760-954
Supplier:   VWR
Supplier #:  J395-400UL
BUFFER AMV REVERSE TRANSCRIPTASE 400UL
Catalog # 80760-954
Supplier:  VWR
CAS Number:  
BUFFER AMV REVERSE TRANSCRIPTASE 400UL
CAS Number:  
Price
$61.86
Log in for availability
Each (400µl)
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Documents

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.