Order Entry
United States
Orders LinkContactUsLinkComponent
Microcide III

Microcide III

5 L
Catalog # 80760-882
Microcide III
Catalog # 80760-882
Supplier:  
CAS Number:  
Microcide III

5 L

CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
MICROCIDE III 1L Catalog #80760-878

Product 80760-878 is similar to 80760-882 but manufacturer and specifications may vary.

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Microcide compound for broad-spectrum antimicrobial activity against bacteria and fungi and are compatible with most formulations.

Synonyms:  5-chloro-2-methyl-1,2-thiazol-3-one
  • Storage Temperature:Ambient

Documents

Specifications

Specifications

  • Size:
    5 L

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.