Order Entry
United States
Orders LinkContactUsLinkComponent
AMP BUFFER CONCENTRATE 20L

AMP BUFFER CONCENTRATE 20L

Catalog # 80760-338
Supplier:   VWR
Supplier #:  BRGT136-20L
AMP BUFFER CONCENTRATE 20L
Catalog # 80760-338
Supplier:  VWR
CAS Number:  
AMP BUFFER CONCENTRATE 20L
CAS Number:  
Price
$1,276.12
Log in for availability
Each (20l)
Product Details & Documents

Documents

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.