PAB RAB 5HYDROXYMETHYLCYTOSINE 25UG/25UL
Catalog # 76183-084
Supplier: Zymo Research
PAB RAB 5HYDROXYMETHYLCYTOSINE 25UG/25UL
Catalog # 76183-084
Supplier: Zymo Research
Supplier Number: A4001-25
Frequently Bought Together
Corning® DMEM (Dulbecco's Modification of Eagle's Medium), Corning
Supplier: Corning
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Weighing Paper
Supplier: VWR International
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Tissue-Tek® O.C.T. Compound, Sakura® Finetek
Supplier: Sakura Finetek
VWR® Media/Storage Bottles with GL Screw Caps
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
Dulbecco's phosphate-buffered saline (DPBS), Mediatech
Supplier: Corning
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Universal Pipette Tips
Supplier: VWR International
Proteinase K
Supplier: New England Biolabs (NEB)
ANTI-DNA HRP ANTIBODY 100X 100UL ELISA
Supplier: Zymo Research