Glutaraldehyde solution 50% in aqueous solution
Glutaraldehyde is a bifunctional protein cross-linking reagent, reacting with NH2 groups to form Schiff's bases.
Sporicidal agent. Glutaraldehyde reacts through cross-linking to impart water resistance to protein and polyhydroxy compounds. It is also a reducing agent for photochemicals. In organic syntheses, the reactive carbonyl groups of glutaraldehyde suggest its use as an intermediate for the production of resins, dyestuffs, and pharmaceuticals. Glutaraldehyde is a disinfectant, which is rapidly effective against vegetative forms of Gram-positive and Gram-negative bacteria. It is also effective against acid-fast bacteria, bacterial spores, some fungi and viruses, including hepatitis B virus and human immunodeficiency virus. It can also be used for stabilization of proteins on agarose beads, activation of polystyrene and glass for immobilization of antibodies and antigens, and coupling peptides onto carrier proteins. Can be used for preparing emulsions for making photographic film. Also for use as an electron microscopy fixative or cell fixation.
Store at +4 °C. Store Under Nitrogen
- Concentration: 50% in H2O
- Viscosity: 105.0 cps (0 °C); 22.1 cps (20 °C); 5.7 cps (40 °C) (Lit.)
- Auto Ignition: 225 °C (DIN 51794)(Lit.)
- Vapor Density: 3.4 (air = 1) (Lit.)
- Vapor Pressure: 22 hPa at 20 °C, 17 mm Hg at 20 °C (Lit.)
- Presentation: Clear Liquid
- :111-30-8
- :C₅H₈O₂
- :MFCD00007025
- :100.12
- :187...189 °C (lit.)
- :1.061 g/cm³ (20 °C) (lit.)
- :Refrigerator
- :2922
- :8,II
- :Density: 1.014 g/cm³ (20 °C)
- Identity Test:Passes
- Specific Gravity @ 20/20°C:1.126-1.135
- Glutaraldehyde Assay:50%-52% by weight
- pH @ 25°C:3.7-4.5
- Dilution Haze Test:1.0 Transmittance Max
- Acidity as HOAC:<40mL KOH in 0.10N methanolic/60.0g
- Heavy Metals:<0.001%
- Pack type:Glass bottle, amber
- Size:100 ml
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



