BIOSENSORS ANTI-HUMAN IGG QUANT. AHQ PK5
Catalog # MSPP-18-5004
Supplier: SARTORIUS CORPORATION MS
BIOSENSORS ANTI-HUMAN IGG QUANT. AHQ PK5
Catalog # MSPP-18-5004
Supplier: SARTORIUS CORPORATION MS
Supplier Number: 18-5004
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
Exel International Brand Quality Luer Lock Syringes, Air-Tite Products
Supplier: Air-Tite
VWR® Razor Blades
Supplier: VWR International
Parafilm® M Sealing Film
Supplier: Bemis Company, Inc.
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
VWR® Upright Freezer Rack with Drawers for 2" Boxes
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Nitrile Examination Gloves, Black
Supplier: VWR International
Reagent alcohol ACS (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Vacuum Filtration Systems featuring Pall Membranes
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Biacore consumables and accessories, Cytiva
Supplier: Cytiva