Orders LinkContactUsLinkComponent
Ward’s® Adjustable Volume Micropipettes
Ward’s® Adjustable Volume Micropipettes
20 μL Micropipette, 2 - 20 µl, 0.5 µl
Catalog # 470231-608
Supplier #:  7010301005
Ward’s® Adjustable Volume Micropipettes
Catalog # 470231-608
Ward’s® Adjustable Volume Micropipettes
Ward’s® Adjustable Volume Micropipettes
20 μL Micropipette, 2 - 20 µl, 0.5 µl
Price
$129.00
Log in for availability
Each Box of 50
Product Details & Documents
The Wards® Micropipette line is fully autoclavable, helping to prevent cross contamination. The ergonomic design is ambidextrous, providing all users with an excellent operating experience.
  • Fully autoclavable
  • Slim, lightweight ergonomic design
  • Compatible with most universal tip brands
  • Tool included for quick and easy recalibration
  • Purchase individually, or as one of our available sets
Personal Pipette Set comes with one of each pipette shown, plus a storage carousel.
Classroom Pipette Set comes with eight of each pipette shown, plus eight storage carousels.
Specifications
  • Volume:
    2 - 20 µl
  • Description:
    20 μL Micropipette
  • Increments:
    0.5 µl