BUFFER DNA/RNA WASH BUFFER CONC. 6ML
Catalog # 77001-874
Supplier: Zymo Research
BUFFER DNA/RNA WASH BUFFER CONC. 6ML
Catalog # 77001-874
Supplier: Zymo Research
Supplier Number: D7010-3-6
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
Hexane (mixture of isomers) ACS, VWR Chemicals BDH®
Supplier: VWR International
Sodium chloride 5 M, sterile-filtered
Supplier: Teknova
Western blotting detection reagent, Amersham™ ECL™ Prime
Supplier: Cytiva
VWR® Sleeve Stoppers
Supplier: VWR International
VWR® Premium Aluminum Foil
Supplier: VWR International
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Talon® Lab Frame Feet
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
RNA ScreenTape Analysis
Supplier: Agilent Technologies
Low Form Zirconium Crucibles, Thermo Scientific
Supplier: Thermo Scientific Chemicals
SYBR® Safe DNA Stain, 10,000 X, Edvotek
Supplier: Edvotek
Transformation of <i>Escherichia coli</i> Lab, Edvotek
Supplier: Edvotek