BUFFER DNA/RNA BINDING BUFFER 10ML
Catalog # 77001-858
Supplier: Zymo Research
BUFFER DNA/RNA BINDING BUFFER 10ML
Catalog # 77001-858
Supplier: Zymo Research
Supplier Number: D7010-1-10
Frequently Bought Together
ROCK SALT
Supplier: INDUSTRIAL SUPPORT INC.
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Nalgene® Invitro™ Biotainer Bottles, Sterile, HDPE, Thermo Scientific
Supplier: Thermo Fisher Scientific
VWR® Talon® Lab Frame Feet
Supplier: VWR International
Benchtop DNA Marker
Supplier: PROMEGA SUNNYVALE INC SE
Ward's® Introduction to Agarose Gel Electrophoresis Lab Activity
Supplier: Avantor
EDVOTEK HexagelTM M36 Electrophoresis Chamber
Supplier: Edvotek
Transformation of <i>Escherichia coli</i> Lab, Edvotek
Supplier: Edvotek
Lok-Tight™ Contact Plates, Hardy Diagnostics
Supplier: Hardy Diagnostics
Low Form Zirconium Crucibles, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Zymit® Low-foam Enzyme Cleaner, International Products
Supplier: International Products
Ward's® Improved Bacterial Transformation Using GFP Lab Activity
Supplier: Avantor
MAGNA AMP LOG 6 PK50
Supplier: MESA LABS MS
INDICATOR CHEM STEAM PK=BX250
Supplier: Steris
SYBR® Safe DNA Stain, 10,000 X, Edvotek
Supplier: Edvotek
Firefly Gene Transformation Kit
Supplier: Avantor