BUFFER CHIP DNA BINDING 50ML GDNA CLEAN
Catalog # 77001-704
Supplier: Zymo Research
BUFFER CHIP DNA BINDING 50ML GDNA CLEAN
Catalog # 77001-704
Supplier: Zymo Research
Supplier Number: D5201-1-50
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Graduated Cylinders, Calibrated To Deliver, Class B
Supplier: VWR International
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
Corning® Internal Thread Cryogenic Vials, Corning
Supplier: Corning
VWR® Razor Blades
Supplier: VWR International
Corning® DMEM (Dulbecco's Modification of Eagle's Medium), Corning
Supplier: Corning
uTIP™ Filtered Pipette Tips for Universal Pipettors, Biotix
Supplier: Biotix
Chloroform ≥99.8% (by GC, corrected for water content) stabilised, BAKER ANALYZED® HPLC for HPLC, for UV spectrophotometry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Petri Dishes
Supplier: VWR International
Cell Culture Storage Bottles, Polystyrene, Corning
Supplier: Corning
VWR® Nitrile Examination Gloves, Black
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Zymo-Spin™ IC-XL Columns, Zymo Research
Supplier: Zymo Research