Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BUFFER M-DESULPHONATION 40 ML EZ DNA KIT
Catalog # 77001-556
Supplier:
Zymo Research
Expand image
BUFFER M-DESULPHONATION 40 ML EZ DNA KIT
Catalog # 77001-556
Supplier:
Zymo Research
Supplier Number:
D5002-5
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Multiwell Cell Culture Plates
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Disposable Transfer Pipettes
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
BUFFER M-BINDING 80 ML F/METHYLATION KIT
Supplier: Zymo Research
Penicillin : Streptomycin solution 50X, Corning®
Supplier: Corning