Order Entry
United States
Orders LinkContactUsLinkComponent
Thread Adapters, Polypropylene

Thread Adapters, Polypropylene

Internal thread: 51 mm (2.01") - 2" BSP, Green
Catalog # 76077-140
Supplier:   BURKLE INC
Supplier #:  0600-2005
Thread Adapters, Polypropylene
Catalog # 76077-140
Supplier:  BURKLE INC
Thread Adapters, Polypropylene
Thread Adapters, Polypropylene

Internal thread: 51 mm (2.01") - 2" BSP, Green

Price
$57.92
Log in for availability
Each
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Bürkle offers a broad range of thread adapters to facilitate secure connections between many Bürkle pumps and discharge cocks to a variety of containers

This wide variety of adapters ensures users can find the correct connection.

Specifications
  • Color:
    Green
  • Description:
    Internal thread: 51 mm (2.01") - 2" BSP
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.